missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)pyrimidine hydrochloride, 97%

Catalog No. AAH32582ME
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32582ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH32582ME Supplier Thermo Scientific Chemicals Supplier No. H32582ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1138011-17-2 | C5H7N3 | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1138011-17-2
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.13
MDL Number MFCD06796202,MFCD07375466
InChI Key AUHXBTKGPUVFCB-UHFFFAOYSA-N
Synonym 4-aminomethyl pyrimidine, 4-pyrimidinemethanamine, 1-pyrimidin-4-yl methanamine, 4-aminomethylpyrimidine, pyrimidin-4-ylmethyl amine dihydrochloride, pyrimidin-4-yl methanamine, c-pyrimidin-4-yl-methylamine, 4-pyrimidinemethanamine 9ci, 1-pyrimidin-4-ylmethanamine dihydrochloride
PubChem CID 12933348
IUPAC Name pyrimidin-4-ylmethanamine
SMILES NCC1=NC=NC=C1

Specifications

Melting Point 176°C to 178°C
Quantity 500 mg
Formula Weight 145.59
Percent Purity 97%
Chemical Name or Material 4-(Aminomethyl)pyrimidine hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only