missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)piperidine 98.0+%, TCI America™

Catalog No. a033125g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
A033125G 25 g
A0331500G 500 g
2 options

Catalog No. A033125G

Supplier: TCI America A033125G

Only null left

Chemical Identifiers

CAS 7144-05-0
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00006007
InChI Key LTEKQAPRXFBRNN-UHFFFAOYSA-N
Synonym 4-aminomethyl piperidine, 4-piperidinemethanamine, 4-aminomethylpiperidine, 4-piperidylmethylamine, 4-amionomethyl piperidine, piperidine, 4-aminomethyl, piperidin-4-yl methanamine, 1-piperidin-4-ylmethanamine, 1-piperidin-4-yl methanamine
PubChem CID 23527
IUPAC Name piperidin-4-ylmethanamine
SMILES C1CNCCC1CN

Specifications

Color White
Boiling Point 92°C
Quantity 25 g
UN Number 3259
Formula Weight 114.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-(Aminomethyl)piperidine