missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)piperidine 98.0+%, TCI America™

Catalog No. A0331500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0331500G 500 g
A033125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0331500G Supplier TCI America Supplier No. A0331500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7144-05-0
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00006007
InChI Key LTEKQAPRXFBRNN-UHFFFAOYSA-N
Synonym 4-aminomethyl piperidine, 4-piperidinemethanamine, 4-aminomethylpiperidine, 4-piperidylmethylamine, 4-amionomethyl piperidine, piperidine, 4-aminomethyl, piperidin-4-yl methanamine, 1-piperidin-4-ylmethanamine, 1-piperidin-4-yl methanamine
PubChem CID 23527
IUPAC Name piperidin-4-ylmethanamine
SMILES C1CNCCC1CN

Specifications

Color White
Boiling Point 92°C
Quantity 500 g
UN Number 3259
Formula Weight 114.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-(Aminomethyl)piperidine