Learn More
4-(Aminomethyl)indole, 97%
CAS: 3468-18-6 | C9H10N2 | 146.19 g/mol
$68.85 - $208.30
Chemical Identifiers
| CAS | 3468-18-6 |
|---|---|
| Molecular Formula | C9H10N2 |
| Molecular Weight (g/mol) | 146.19 |
| MDL Number | MFCD04973297 |
| InChI Key | FFBWKPKOXRMLNP-UHFFFAOYSA-N |
| Synonym | 4-aminomethylindole, 1h-indol-4-yl methanamine, 1h-indole-4-methanamine, 4-aminomethyl indole, 1h-indol-4-ylmethyl amine, 1-1h-indol-4-yl methanamine, c-1h-indol-4-yl-methylamine, indole-4-methylamine, 4-aminomethyl-indole, 4-aminomethyl-1h-indole |
| PubChem CID | 280302 |
| IUPAC Name | 1H-indol-4-ylmethanamine |
| SMILES | NCC1=C2C=CNC2=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH54963MD
|
Thermo Scientific Chemicals
H54963MD |
250 mg |
Each for $68.85
|
|
|||||
|
AAH5496303
|
Thermo Scientific Chemicals
H5496303 |
1 g |
Each for $208.30
|
|
|||||
Description
4-(Aminomethyl)indole is used as a reactant for preparation of dopamine receptor antagonists and for preparation of high-affinity ligands to α2δ subunit of voltage-gated calcium channels. It is used as a synthetic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Aminomethyl)indole is used as a reactant for preparation of dopamine receptor antagonists and for preparation of high-affinity ligands to α2δ subunit of voltage-gated calcium channels. It is used as a synthetic intermediate.
Solubility
Soluble in ethanol, dimethyl sulfoxide and dimethyl formamide.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
| 3468-18-6 | |
| 146.19 | |
| FFBWKPKOXRMLNP-UHFFFAOYSA-N | |
| 280302 | |
| NCC1=C2C=CNC2=CC=C1 |
| C9H10N2 | |
| MFCD04973297 | |
| 4-aminomethylindole, 1h-indol-4-yl methanamine, 1h-indole-4-methanamine, 4-aminomethyl indole, 1h-indol-4-ylmethyl amine, 1-1h-indol-4-yl methanamine, c-1h-indol-4-yl-methylamine, indole-4-methylamine, 4-aminomethyl-indole, 4-aminomethyl-1h-indole | |
| 1H-indol-4-ylmethanamine |
Specifications
| 3468-18-6 | |
| C9H10N2 | |
| 250 mg | |
| Soluble in ethanol,dimethyl sulfoxide and dimethyl formamide. | |
| NCC1=C2C=CNC2=CC=C1 | |
| 146.19 | |
| 146.19 | |
| 4-(Aminomethyl)indole |
| 130°C to 135°C | |
| MFCD04973297 | |
| 4-aminomethylindole, 1h-indol-4-yl methanamine, 1h-indole-4-methanamine, 4-aminomethyl indole, 1h-indol-4-ylmethyl amine, 1-1h-indol-4-yl methanamine, c-1h-indol-4-yl-methylamine, indole-4-methylamine, 4-aminomethyl-indole, 4-aminomethyl-1h-indole | |
| FFBWKPKOXRMLNP-UHFFFAOYSA-N | |
| 1H-indol-4-ylmethanamine | |
| 280302 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only