missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)-1-tert-butoxycarbonylpiperidine 98.0+%, TCI America™

Catalog No. B44455G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44455G 5 g
B44451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44455G Supplier TCI America Supplier No. B44455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 144222-22-0
Molecular Formula C11H23N2O2
Molecular Weight (g/mol) 215.32
MDL Number MFCD01076207
InChI Key KLKBCNDBOVRQIJ-UHFFFAOYSA-O
Synonym 1-boc-4-aminomethyl piperidine, tert-butyl 4-aminomethyl piperidine-1-carboxylate, 1-boc-4-aminomethylpiperidine, 1-boc-4-aminomethyl-piperidine, 4-aminomethyl-1-n-boc-piperidine, boc-4-aminomethyl-piperidine, 4-aminomethyl-1-n-t-butoxycarbonyl piperidine, 4-aminomethyl-1-boc-piperidine, 1-n-boc-4-aminomethyl piperidine, 4-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester
PubChem CID 736817
IUPAC Name {1-[(tert-butoxy)carbonyl]piperidin-4-yl}methanaminium
SMILES CC(C)(C)OC(=O)N1CCC(C[NH3+])CC1

Specifications

Color Yellow
Boiling Point 238°C
Quantity 5 g
Formula Weight 214.31
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(Aminomethyl)-1-tert-butoxycarbonylpiperidine