missing translation for 'onlineSavingsMsg'
Learn More

4-Aminomethyl-1-Boc-piperidine, 97%

Catalog No. p-7035111
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5281906 5 g
AAH5281914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5281906 Supplier Thermo Scientific Chemicals Supplier No. H5281906
Only null left

CAS: 144222-22-0 | C11H23N2O2 | 215.32 g/mol

4-Aminomethyl-1-Boc-piperidine is used in the preparation biologically active compounds and orally active platelet-activating factor antagonists. It is used as an inhibitor for aspartic acid protease and Kinesin spindle protein. It is an active reactant involved in the enantioselective synthesis of N-alkyl terminal aziridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminomethyl-1-Boc-piperidine is used in the preparation biologically active compounds and orally active platelet-activating factor antagonists. It is used as an inhibitor for aspartic acid protease and Kinesin spindle protein. It is an active reactant involved in the enantioselective synthesis of N-alkyl terminal aziridines.

Solubility
Soluble in chloroform and methanol.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 144222-22-0
Molecular Formula C11H23N2O2
Molecular Weight (g/mol) 215.32
MDL Number MFCD01076207
InChI Key KLKBCNDBOVRQIJ-UHFFFAOYSA-O
Synonym 1-boc-4-aminomethyl piperidine, tert-butyl 4-aminomethyl piperidine-1-carboxylate, 1-boc-4-aminomethylpiperidine, 1-boc-4-aminomethyl-piperidine, 4-aminomethyl-1-n-boc-piperidine, boc-4-aminomethyl-piperidine, 4-aminomethyl-1-n-t-butoxycarbonyl piperidine, 4-aminomethyl-1-boc-piperidine, 1-n-boc-4-aminomethyl piperidine, 4-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester
PubChem CID 736817
IUPAC Name {1-[(tert-butoxy)carbonyl]piperidin-4-yl}methanaminium
SMILES CC(C)(C)OC(=O)N1CCC(C[NH3+])CC1

Specifications

Density 1.013
Boiling Point 237°C to 238°C
Flash Point >110°C (230°F)
Quantity 5 g
Solubility Information Soluble in chloroform and methanol.
Formula Weight 214.31
Percent Purity 97%
Chemical Name or Material 4-Aminomethyl-1-Boc-piperidine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only