missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminoisoquinoline 98.0+%, TCI America™
$83.89 - $331.81
Chemical Identifiers
| CAS | 23687-25-4 |
|---|---|
| Molecular Formula | C9H8N2 |
| Molecular Weight (g/mol) | 144.177 |
| MDL Number | MFCD00034752 |
| InChI Key | ISIUXVGHQFJYHM-UHFFFAOYSA-N |
| PubChem CID | 90237 |
| IUPAC Name | isoquinolin-4-amine |
| SMILES | C1=CC=C2C(=C1)C=NC=C2N |
Chemical Identifiers
| 23687-25-4 | |
| 144.177 | |
| ISIUXVGHQFJYHM-UHFFFAOYSA-N | |
| isoquinolin-4-amine |
| C9H8N2 | |
| MFCD00034752 | |
| 90237 | |
| C1=CC=C2C(=C1)C=NC=C2N |
Specifications
| 23687-25-4 | |
| C9H8N2 | |
| 1 g | |
| C1=CC=C2C(=C1)C=NC=C2N | |
| 144.177 | |
| 144.18 | |
| Crystalline Powder |
| 110°C | |
| MFCD00034752 | |
| ISIUXVGHQFJYHM-UHFFFAOYSA-N | |
| isoquinolin-4-amine | |
| 90237 | |
| ≥98.0% (GC,T) | |
| 4-Aminoisoquinoline |
Safety and Handling
TSCA : No