missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoindole 98.0+%, TCI America™

Catalog No. p-7117712
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20701G 1 g
A20705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20701G Supplier TCI America Supplier No. A20701G

Chemical Identifiers

CAS 5192-23-4
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.166
MDL Number MFCD01076559
InChI Key LUNUNJFSHKSXGQ-UHFFFAOYSA-N
Synonym 4-aminoindole, 1h-indol-4-ylamine, 4-amino indole, 4-amino-1h-indole, 1h-indol-4-yl-amine, 4-indolamine, indole-4-ylamine, 1h-indol-4-amin, 4-aminoindol, indol-4-ylamine
PubChem CID 583431
IUPAC Name 1H-indol-4-amine
SMILES C1=CC2=C(C=CN2)C(=C1)N

Specifications

Melting Point 109°C
Quantity 1 g
Formula Weight 132.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminoindole