missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoimidazole-5-carboxamide hydrochloride, 98%, Thermo Scientific Chemicals

Catalog No. AAB2401206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2401206 5 g
AAB2401214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2401206 Supplier Thermo Scientific Chemicals Supplier No. B2401206

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 72-40-2
Molecular Formula C4H7ClN4O
Molecular Weight (g/mol) 162.58
MDL Number MFCD00012704
InChI Key MXCUYSMIELHIQL-UHFFFAOYSA-N
Synonym 4-amino-5-imidazolecarboxamide hydrochloride, 5-amino-4-imidazolecarboxamide hydrochloride, 5-amino-1h-imidazole-4-carboxamide hydrochloride, 5-aminoimidazole-4-carboxamide hydrochloride, aic .cntdot. hcl, 4-amino-1h-imidazole-5-carboxamide hydrochloride, aminoimidazolecarboxamide hydrochloride, 4-aminoimidazole-5-carboxamide hydrochloride, 1h-imidazole-4-carboxamide, 5-amino-, monohydrochloride, 5-aminoimidazol-4-carboxamide, hydrochloride
PubChem CID 66146
IUPAC Name 4-amino-1H-imidazole-5-carboxamide;hydrochloride
SMILES Cl.NC(=O)C1=C(N)N=CN1

Specifications

Melting Point ∼270°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 3701645
Sensitivity Hygroscopic
Formula Weight 162.58
Percent Purity 98%
Chemical Name or Material 4-Aminoimidazole-5-carboxamide hydrochloride
EINECSNumber 200-778-3
RTECSNumber NI3911000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only