missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoimidazole-5-carboxamide, 95%

Catalog No. p-7056941
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6618406 5 g
AAH6618414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6618406 Supplier Thermo Scientific Chemicals Supplier No. H6618406

CAS: 360-97-4 | C4H6N4O | 126.12 g/mol

4-Aminoimidazole-5-carboxamide is used as Catalytic agent, Petrochemical additive. It is metabolite of temozolomide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminoimidazole-5-carboxamide is used as Catalytic agent, Petrochemical additive. It is metabolite of temozolomide.

Solubility
Soluble in DMSO, Methanol, Water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 360-97-4
Molecular Formula C4H6N4O
Molecular Weight (g/mol) 126.12
MDL Number MFCD02181040
InChI Key DVNYTAVYBRSTGK-UHFFFAOYSA-N
Synonym 5-aminoimidazole-4-carboxamide, 5-amino-1h-imidazole-4-carboxamide, 4-amino-5-imidazolecarboxamide, 5-amino-4-imidazolecarboxamide, colahepat, 4-aminoimidazole-5-carboxamide, aica, 1h-imidazole-4-carboxamide, 5-amino, diazol-c, 4-carbamoyl-5-aminoimidazole
PubChem CID 9679
ChEBI CHEBI:2030
IUPAC Name 4-amino-1H-imidazole-5-carboxamide
SMILES NC(=O)C1=C(N)N=CN1

Specifications

Melting Point 164°C to 170°C
Color White
Quantity 5 g
Beilstein 3938
Solubility Information Soluble in DMSO,Methanol,Water.
Formula Weight 126.12
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Aminoimidazole-5-carboxamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-641-4
RTECSNumber NI3910000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only