missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminohydrocinnamic Acid 98.0+%, TCI America™
$247.92 - $480.92
Chemical Identifiers
| CAS | 2393-17-1 |
|---|---|
| Molecular Formula | C9H11NO2 |
| Molecular Weight (g/mol) | 165.192 |
| MDL Number | MFCD00017118 |
| InChI Key | WXOHKMNWMKZMND-UHFFFAOYSA-N |
| Synonym | 3-4-aminophenyl propionic acid, 3-4-aminophenyl propanoic acid, 4-aminohydrocinnamic acid, benzenepropanoic acid, 4-amino, 3-p-aminophenyl propionic acid, beta-p-aminophenyl propionic acid, p-aminohydrocinnamic acid, 4-aminobenzenepropanoic acid, benzenepropanoic acid,4-amino, beta-4-aminophenyl propionic acid |
| PubChem CID | 75451 |
| IUPAC Name | 3-(4-aminophenyl)propanoic acid |
| SMILES | C1=CC(=CC=C1CCC(=O)O)N |
Chemical Identifiers
| 2393-17-1 | |
| 165.192 | |
| WXOHKMNWMKZMND-UHFFFAOYSA-N | |
| 75451 | |
| C1=CC(=CC=C1CCC(=O)O)N |
| C9H11NO2 | |
| MFCD00017118 | |
| 3-4-aminophenyl propionic acid, 3-4-aminophenyl propanoic acid, 4-aminohydrocinnamic acid, benzenepropanoic acid, 4-amino, 3-p-aminophenyl propionic acid, beta-p-aminophenyl propionic acid, p-aminohydrocinnamic acid, 4-aminobenzenepropanoic acid, benzenepropanoic acid,4-amino, beta-4-aminophenyl propionic acid | |
| 3-(4-aminophenyl)propanoic acid |
Specifications
| 2393-17-1 | |
| C9H11NO2 | |
| 10 g | |
| WXOHKMNWMKZMND-UHFFFAOYSA-N | |
| 3-(4-aminophenyl)propanoic acid | |
| 75451 | |
| ≥98.0% (GC,T) | |
| 4-Aminohydrocinnamic Acid |
| 132°C | |
| MFCD00017118 | |
| 3-4-aminophenyl propionic acid, 3-4-aminophenyl propanoic acid, 4-aminohydrocinnamic acid, benzenepropanoic acid, 4-amino, 3-p-aminophenyl propionic acid, beta-p-aminophenyl propionic acid, p-aminohydrocinnamic acid, 4-aminobenzenepropanoic acid, benzenepropanoic acid,4-amino, beta-4-aminophenyl propionic acid | |
| C1=CC(=CC=C1CCC(=O)O)N | |
| 165.192 | |
| 165.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No