missing translation for 'onlineSavingsMsg'
Learn More

4-Aminohydrocinnamic Acid 98.0+%, TCI America™

Catalog No. A107325G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A107325G 25 g
A107310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A107325G Supplier TCI America Supplier No. A107325G
Only null left

Chemical Identifiers

CAS 2393-17-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00017118
InChI Key WXOHKMNWMKZMND-UHFFFAOYSA-N
Synonym 3-4-aminophenyl propionic acid, 3-4-aminophenyl propanoic acid, 4-aminohydrocinnamic acid, benzenepropanoic acid, 4-amino, 3-p-aminophenyl propionic acid, beta-p-aminophenyl propionic acid, p-aminohydrocinnamic acid, 4-aminobenzenepropanoic acid, benzenepropanoic acid,4-amino, beta-4-aminophenyl propionic acid
PubChem CID 75451
IUPAC Name 3-(4-aminophenyl)propanoic acid
SMILES C1=CC(=CC=C1CCC(=O)O)N

Specifications

Melting Point 132°C
Quantity 25 g
Formula Weight 165.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminohydrocinnamic Acid