missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminocyclohexanecarboxylic Acid (cis- and trans- mixture) 95.0+%, TCI America™
$150.98 - $447.38
Chemical Identifiers
| CAS | 1776-53-0 |
|---|---|
| Molecular Formula | C7H13NO2 |
| Molecular Weight (g/mol) | 143.186 |
| MDL Number | MFCD00191601 |
| InChI Key | DRNGLYHKYPNTEA-UHFFFAOYSA-N |
| Synonym | 4-Aminohexahydrobenzoic Acid |
| PubChem CID | 171560 |
| IUPAC Name | 4-aminocyclohexane-1-carboxylic acid |
| SMILES | C1CC(CCC1C(=O)O)N |
Chemical Identifiers
| 1776-53-0 | |
| 143.186 | |
| DRNGLYHKYPNTEA-UHFFFAOYSA-N | |
| 171560 | |
| C1CC(CCC1C(=O)O)N |
| C7H13NO2 | |
| MFCD00191601 | |
| 4-Aminohexahydrobenzoic Acid | |
| 4-aminocyclohexane-1-carboxylic acid |
Specifications
| 1776-53-0 | |
| C7H13NO2 | |
| 4-Aminohexahydrobenzoic Acid | |
| C1CC(CCC1C(=O)O)N | |
| 5 g | |
| 171560 | |
| ≥95.0% (GC,T) | |
| 4-Aminocyclohexanecarboxylic Acid (cis- and trans- mixture) |
| White-Yellow | |
| MFCD00191601 | |
| DRNGLYHKYPNTEA-UHFFFAOYSA-N | |
| 4-aminocyclohexane-1-carboxylic acid | |
| 143.186 | |
| 143.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No