missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobutyric acid, 99+%

Catalog No. AAJ6130709
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6130709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6130709 Supplier Thermo Scientific Chemicals Supplier No. J6130709

CAS: 56-12-2 | C4H9NO2 | 103.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56-12-2
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00008226
InChI Key BTCSSZJGUNDROE-UHFFFAOYSA-N
Synonym 4-aminobutyric acid, gaba, gamma-aminobutyric acid, piperidic acid, piperidinic acid, aminalon, gaballon, gammalon, mielogen, butanoic acid, 4-amino
PubChem CID 119
ChEBI CHEBI:16865
IUPAC Name 4-aminobutanoic acid
SMILES C(CC(=O)O)CN

Specifications

Melting Point ∽195°C (decomposition)
Color White
Linear Formula H2N(CH2)3CO2H
Beilstein 906818
Merck Index 14,430
Quantity 10 g
Odor Odorless
Formula Weight 103.12
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material 4-Aminobutyric acid
Show More Show Less
EINECSNumber 200-258-6
RTECSNumber ES6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only