missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobutyric acid, 99+%, Thermo Scientific™

4-Aminobutyric acid, 99+%, CAS # 56-12-2, also known as γ-Aminobutyric acid, is an amino acid that shows inhibitory activities against neurotransmitters, including GABA.

Supplier:  Thermo Scientific Chemicals 103280025

Catalog No. AC103280025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

• 4-Aminobutyric acid is a γ-amino acid and monocarboxylic acid derived from butyric acid

• It is an amino acid that shows inhibitory activities against neurotransmitters, including GABA, and acts by binding GABA-A receptor and inhibiting the activity of signal-receiving neurons

• This compound, converted from neurotransmitter glutamate, is essential to regulate neuronal excitability

• It binds to GABA-A and GABA-B receptors, causing ion channel opening, hyperpolarization, and eventually inhibition of neurotransmission

Applications

• 4-Aminobutyric acid has been used in the preparation of GABA–BSA (bovine serum albumin)–GA (glutaraldehyde) conjugate, which is used as a control for immunohistochemical experiments

• It is used, in vivo, to detect GABA neurotransmitter by magnetic resonance spectroscopy

• This compound increases the production of short-chain fatty acids and decreases pH values in mouse colon

• Some in vivo experiments, using rats, mice, and pigs, demonstrated that 4-Aminobutyric acid can modulate intestinal fluid secretion, blood flow and ulceration

Specifications

Specifications

4-Aminobutyric acid
56-12-2
100.0
Authentic
Plastic bottle
H2N(CH2)3CO2H
2.5kg
15,425
Solubility in water: very soluble. Other solubilities: slightly soluble in common solvents
C(CC(=O)O)CN
95% min. (c=10, 2N HCl, 430nm) 1cm cell
119
103.12
99+%
99.0
0.5% max.
99+%
C4H9NO2
MFCD00008226
04,413
4-aminobutyric acid, gaba, gamma-aminobutyric acid, piperidic acid, piperidinic acid, aminalon, gaballon, gammalon, mielogen, butanoic acid, 4-amino
BTCSSZJGUNDROE-UHFFFAOYSA-N
4-aminobutanoic acid
103.12
CHEBI:16865
99+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-258-6

RTECSNumber : ES6300000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only