missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobutyric acid, 99+%

Catalog No. AC103285000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103285000 500 g Plastic Bottle
AC103280250 25 g Plastic Bottle
AC103281000 100 g Plastic Bottle
AC103280025 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC103285000 Supplier Thermo Scientific Chemicals Supplier No. 103285000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

4-Aminobutyric acid, 99+%, CAS # 56-12-2, also known as γ-Aminobutyric acid, is an amino acid that shows inhibitory activities against neurotransmitters, including GABA. | CAS: 56-12-2 | C4H9NO2 | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• 4-Aminobutyric acid is a γ-amino acid and monocarboxylic acid derived from butyric acid

• It is an amino acid that shows inhibitory activities against neurotransmitters, including GABA, and acts by binding GABA-A receptor and inhibiting the activity of signal-receiving neurons

• This compound, converted from neurotransmitter glutamate, is essential to regulate neuronal excitability

• It binds to GABA-A and GABA-B receptors, causing ion channel opening, hyperpolarization, and eventually inhibition of neurotransmission

Applications

• 4-Aminobutyric acid has been used in the preparation of GABA–BSA (bovine serum albumin)–GA (glutaraldehyde) conjugate, which is used as a control for immunohistochemical experiments

• It is used, in vivo, to detect GABA neurotransmitter by magnetic resonance spectroscopy

• This compound increases the production of short-chain fatty acids and decreases pH values in mouse colon

• Some in vivo experiments, using rats, mice, and pigs, demonstrated that 4-Aminobutyric acid can modulate intestinal fluid secretion, blood flow and ulceration

Chemical Identifiers

CAS 56-12-2
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00008226
InChI Key BTCSSZJGUNDROE-UHFFFAOYSA-N
Synonym 4-aminobutyric acid, gaba, gamma-aminobutyric acid, piperidic acid, piperidinic acid, aminalon, gaballon, gammalon, mielogen, butanoic acid, 4-amino
PubChem CID 119
ChEBI CHEBI:16865
IUPAC Name 4-aminobutanoic acid
SMILES C(CC(=O)O)CN

Specifications

Melting Point 197°C to 204°C
CAS Min % 99.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Loss on Drying 0.5% max.
Assay Percent Range 99+%
Packaging Plastic Bottle
Linear Formula H2N(CH2)3CO2H
Beilstein 04,413
Transmittance 95% min. (c=10, 2N HCl, 430nm) 1cm cell
Merck Index 15,425
Solubility Information Solubility in water: very soluble. Other solubilities: slightly soluble in common solvents
Quantity 500 g
Formula Weight 103.12
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminobutyric acid, 99+%
Show More Show Less
EINECSNumber 200-258-6
RTECSNumber ES6300000
TSCA TSCA

RUO – Research Use Only