missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobutyric acid, 97%

Catalog No. AAA1101622
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1101622 100 g
AAA1101636 500 g
AAA110160E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1101622 Supplier Thermo Scientific Chemicals Supplier No. A1101622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56-12-2 | C4H9NO2 | 103.121 g/mol

In the production of polymers, food additives, and pharmaceuticals. Acrylic acid and methacrylic acids are well known precursors of polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In the production of polymers, food additives, and pharmaceuticals. Acrylic acid and methacrylic acids are well known precursors of polymers.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 56-12-2
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00008226
InChI Key BTCSSZJGUNDROE-UHFFFAOYSA-N
Synonym 4-aminobutyric acid, gaba, gamma-aminobutyric acid, piperidic acid, piperidinic acid, aminalon, gaballon, gammalon, mielogen, butanoic acid, 4-amino
PubChem CID 119
ChEBI CHEBI:16865
IUPAC Name 4-aminobutanoic acid
SMILES C(CC(=O)O)CN

Specifications

Melting Point ∼195°C (decomposition)
Linear Formula H2N(CH2)3CO2H
Beilstein 906818
Merck Index 14,430
Solubility Information Soluble in water.
Quantity 100 g
Odor Odorless
Formula Weight 103.12
Percent Purity 97%
Chemical Name or Material 4-Aminobutyric acid
EINECSNumber 200-258-6
RTECSNumber ES6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only