Learn More
4-Aminobutyraldehyde dimethyl acetal, 98+%
CAS: 19060-15-2 | C6H16NO2 | 134.20 g/mol
$102.92 - $361.89
Chemical Identifiers
| CAS | 19060-15-2 |
|---|---|
| Molecular Formula | C6H16NO2 |
| Molecular Weight (g/mol) | 134.20 |
| MDL Number | MFCD00189373 |
| InChI Key | TYVAXMOICMBSMT-UHFFFAOYSA-O |
| Synonym | 4-aminobutyraldehyde dimethyl acetal, 4,4-dimethoxybutylamine, 4,4-dimethoxy-1-butanamine, 4-aminobutyraldehyde dimethylacetal, acmc-1bqj6, 4-aminobutyraldehyde dimethyl, 1-amino-4,4-dimethoxybutane, cs-waa0041, tyvaxmoicmbsmt-uhfffaoysa, 4-aminobutyraldehydedimethylacetal |
| PubChem CID | 5463803 |
| SMILES | COC(CCC[NH3+])OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2107206
|
Thermo Scientific Chemicals
B2107206 |
5 g |
Each for $102.92
|
|
|||||
|
AAB2107214
|
Thermo Scientific Chemicals
B2107214 |
25 g |
Each for $361.89
|
|
|||||
Description
4-Aminobutyraldehyde dimethyl acetal acts as an intermediate in organic synthesis. It is used in the preparation of pineal hormone melatonin by reaction with 4-methoxyphenylhydrazine hydrochloride and acetic anhydride. Further, it is also used in the preparation of ficuseptine, juliprosine, and juliprosopine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Aminobutyraldehyde dimethyl acetal acts as an intermediate in organic synthesis. It is used in the preparation of pineal hormone melatonin by reaction with 4-methoxyphenylhydrazine hydrochloride and acetic anhydride. Further, it is also used in the preparation of ficuseptine, juliprosine, and juliprosopine.
Solubility
Miscible with benzene.
Notes
Air sensitive. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
Chemical Identifiers
| 19060-15-2 | |
| 134.20 | |
| TYVAXMOICMBSMT-UHFFFAOYSA-O | |
| 5463803 |
| C6H16NO2 | |
| MFCD00189373 | |
| 4-aminobutyraldehyde dimethyl acetal, 4,4-dimethoxybutylamine, 4,4-dimethoxy-1-butanamine, 4-aminobutyraldehyde dimethylacetal, acmc-1bqj6, 4-aminobutyraldehyde dimethyl, 1-amino-4,4-dimethoxybutane, cs-waa0041, tyvaxmoicmbsmt-uhfffaoysa, 4-aminobutyraldehydedimethylacetal | |
| COC(CCC[NH3+])OC |
Specifications
| 19060-15-2 | |
| 59°C (138°F) | |
| 1.43 | |
| MFCD00189373 | |
| UN2734 | |
| 4-aminobutyraldehyde dimethyl acetal, 4,4-dimethoxybutylamine, 4,4-dimethoxy-1-butanamine, 4-aminobutyraldehyde dimethylacetal, acmc-1bqj6, 4-aminobutyraldehyde dimethyl, 1-amino-4,4-dimethoxybutane, cs-waa0041, tyvaxmoicmbsmt-uhfffaoysa, 4-aminobutyraldehydedimethylacetal | |
| TYVAXMOICMBSMT-UHFFFAOYSA-O | |
| 4,4-dimethoxybutan-1-aminium | |
| 5463803 | |
| ≥98% |
| 85°C (22 mmHg) | |
| C6H16NO2 | |
| H2N(CH2)3CH(OCH3)2 | |
| 5 g | |
| Air Sensitive | |
| Miscible with benzene. | |
| COC(CCC[NH3+])OC | |
| 134.20 | |
| 133.19 | |
| 4-Aminobutyraldehyde dimethyl acetal |
Safety and Handling
GHS H Statement
H314-H226-H302-H317
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
May cause an allergic skin reaction.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H302-H314-H334-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber : 407-690-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only