missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobutyraldehyde diethyl acetal, 95%

Catalog No. AC103260050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103260050 5 g Glass bottle
AC103260250 25 g Glass bottle
AC103261000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC103260050 Supplier Thermo Scientific Chemicals Supplier No. 103260050
Only null left

CAS: 6346-09-4 | C8H19NO2 | 161.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6346-09-4
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.24
MDL Number MFCD00008227
InChI Key GFLPSABXBDCMCN-UHFFFAOYSA-N
IUPAC Name 4,4-diethoxybutan-1-amine
SMILES CCOC(CCCN)OCC

Specifications

Color Colorless
Density 0.9000g/mL
Boiling Point 196.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4270 to 1.4290
Linear Formula H2N(CH2)3CH(OC2H5)2
Quantity 5 g
Beilstein 04,319
Specific Gravity 0.9
Synonym 4,4-diethoxybutylamine, 4-aminobutyraldehyde diethyl acetal, 1-butanamine, 4,4-diethoxy, 4,4-diethoxy-1-butanamine, ambda, 4,4-diethoxy-butylamine, acmc-2097jf, 4-aminobutanal diethyl acetal, ksc356g0p, 4,4-diethoxy-1-butanamine #
Solubility Information Solubility in water: miscible.
PubChem CID 80672
Formula Weight 161.24
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Aminobutyraldehyde diethyl acetal
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 228-753-2

RUO – Research Use Only