missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzonitrile 98.0+%, TCI America™

Catalog No. p-7117011
Change view
Click to view available options
Quantity:
10 g
250 g
Catalog No. Quantity
A069510G 10 g
A0695250G 250 g
2 options

Catalog No. A069510G

Supplier: TCI America A069510G

Only null left

Chemical Identifiers

CAS 873-74-5
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00007821
InChI Key YBAZINRZQSAIAY-UHFFFAOYSA-N
Synonym 4-cyanoaniline, p-aminobenzonitrile, p-cyanoaniline, benzonitrile, 4-amino, aniline, p-cyano, benzonitrile, p-amino, 1-amino-4-cyanobenzene, unii-pb8v69y76t, 4-amino benzonitrile, 4-amino-benzonitrile
PubChem CID 13396
IUPAC Name 4-aminobenzonitrile
SMILES C1=CC(=CC=C1C#N)N

Specifications

Melting Point 86°C
Boiling Point 167°C
Quantity 10 g
UN Number 3439
Formula Weight 118.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminobenzonitrile
EINECSNumber (3)-1812
RTECSNumber BX2320000
TSCA Yes