Learn More
4-Aminobenzeneboronic acid hydrochloride, 97%
CAS: 80460-73-7 | C6H9BClNO2 | 173.403 g/mol
$163.82 - $450.15
Chemical Identifiers
| CAS | 80460-73-7 |
|---|---|
| Molecular Formula | C6H9BClNO2 |
| Molecular Weight (g/mol) | 173.403 |
| MDL Number | MFCD03001333 |
| InChI Key | QBYGJJSFMOVYOA-UHFFFAOYSA-N |
| Synonym | 4-aminophenylboronic acid hydrochloride, 4-aminophenyl boronic acid hydrochloride, 4-aminobenzeneboronic acid hydrochloride, 4-aminophenylboronic acid hcl, 4-boronoaniline hydrochloride, 4-aminophenylboronic acid, hcl, boronic acid, 4-aminophenyl-, hydrochloride, pubchem1747, 4-aminophenylboronicacidhydrochloride, ksc494e4p |
| PubChem CID | 2734614 |
| IUPAC Name | (4-aminophenyl)boronic acid;hydrochloride |
| SMILES | B(C1=CC=C(C=C1)N)(O)O.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH27479MC
|
Thermo Scientific Chemicals
H27479MC |
100 mg |
Each for $163.82
|
|
|||||
|
AAH27479ME
|
Thermo Scientific Chemicals
H27479ME |
500 mg |
Each for $450.15
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 80460-73-7 | |
| 173.403 | |
| QBYGJJSFMOVYOA-UHFFFAOYSA-N | |
| 2734614 | |
| B(C1=CC=C(C=C1)N)(O)O.Cl |
| C6H9BClNO2 | |
| MFCD03001333 | |
| 4-aminophenylboronic acid hydrochloride, 4-aminophenyl boronic acid hydrochloride, 4-aminobenzeneboronic acid hydrochloride, 4-aminophenylboronic acid hcl, 4-boronoaniline hydrochloride, 4-aminophenylboronic acid, hcl, boronic acid, 4-aminophenyl-, hydrochloride, pubchem1747, 4-aminophenylboronicacidhydrochloride, ksc494e4p | |
| (4-aminophenyl)boronic acid;hydrochloride |
Specifications
| 80460-73-7 | |
| C6H9BClNO2 | |
| 100 mg | |
| 4-aminophenylboronic acid hydrochloride, 4-aminophenyl boronic acid hydrochloride, 4-aminobenzeneboronic acid hydrochloride, 4-aminophenylboronic acid hcl, 4-boronoaniline hydrochloride, 4-aminophenylboronic acid, hcl, boronic acid, 4-aminophenyl-, hydrochloride, pubchem1747, 4-aminophenylboronicacidhydrochloride, ksc494e4p | |
| B(C1=CC=C(C=C1)N)(O)O.Cl | |
| 173.403 | |
| 173.41 | |
| 4-Aminobenzeneboronic acid hydrochloride |
| 125°C to 132°C | |
| MFCD03001333 | |
| Hygroscopic | |
| QBYGJJSFMOVYOA-UHFFFAOYSA-N | |
| (4-aminophenyl)boronic acid;hydrochloride | |
| 2734614 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only