missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzeneboronic acid hydrochloride, 97%

Catalog No. AAH27479MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27479MC 100 mg
AAH27479ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27479MC Supplier Thermo Scientific Chemicals Supplier No. H27479MC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 80460-73-7 | C6H9BClNO2 | 173.403 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80460-73-7
Molecular Formula C6H9BClNO2
Molecular Weight (g/mol) 173.403
MDL Number MFCD03001333
InChI Key QBYGJJSFMOVYOA-UHFFFAOYSA-N
Synonym 4-aminophenylboronic acid hydrochloride, 4-aminophenyl boronic acid hydrochloride, 4-aminobenzeneboronic acid hydrochloride, 4-aminophenylboronic acid hcl, 4-boronoaniline hydrochloride, 4-aminophenylboronic acid, hcl, boronic acid, 4-aminophenyl-, hydrochloride, pubchem1747, 4-aminophenylboronicacidhydrochloride, ksc494e4p
PubChem CID 2734614
IUPAC Name (4-aminophenyl)boronic acid;hydrochloride
SMILES B(C1=CC=C(C=C1)N)(O)O.Cl

Specifications

Melting Point 125°C to 132°C
Quantity 100 mg
Sensitivity Hygroscopic
Formula Weight 173.41
Percent Purity 97%
Chemical Name or Material 4-Aminobenzeneboronic acid hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only