missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzamidoxime, 97%

CAS: 277319-62-7 | C7H9N3O | 151.169 g/mol

$111.58 - $411.32

Chemical Identifiers

CAS 277319-62-7
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.169
MDL Number MFCD08061151
InChI Key CNFNMMJKXWOLPY-UHFFFAOYSA-N
Synonym 4-amino-n'-hydroxybenzimidamide, 4-amino-n'-hydroxybenzene-1-carboximidamide, z-4-amino-n'-hydroxybenzenecarboximidamide
PubChem CID 5478941
IUPAC Name 4-amino-N'-hydroxybenzenecarboximidamide
SMILES C1=CC(=CC=C1C(=NO)N)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5178803
View Documents
Thermo Scientific Chemicals
H5178803
1 g
Each for $111.58
Only null left
 
AAH5178806
View Documents
Thermo Scientific Chemicals
H5178806
5 g
Each for $411.32
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

277319-62-7
151.169
CNFNMMJKXWOLPY-UHFFFAOYSA-N
5478941
C1=CC(=CC=C1C(=NO)N)N
C7H9N3O
MFCD08061151
4-amino-n'-hydroxybenzimidamide, 4-amino-n'-hydroxybenzene-1-carboximidamide, z-4-amino-n'-hydroxybenzenecarboximidamide
4-amino-N'-hydroxybenzenecarboximidamide

Specifications

277319-62-7
C7H9N3O
1 g
CNFNMMJKXWOLPY-UHFFFAOYSA-N
4-amino-N'-hydroxybenzenecarboximidamide
5478941
97%
168°C to 170°C
MFCD08061151
4-amino-n'-hydroxybenzimidamide, 4-amino-n'-hydroxybenzene-1-carboximidamide, z-4-amino-n'-hydroxybenzenecarboximidamide
C1=CC(=CC=C1C(=NO)N)N
151.169
151.17
4-Aminobenzamidoxime
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only