missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzamidoxime, 97%

Catalog No. AAH5178803
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5178803 1 g
AAH5178806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. AAH5178803 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' H5178803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 277319-62-7 | C7H9N3O | 151.169 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 277319-62-7
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.169
MDL Number MFCD08061151
InChI Key CNFNMMJKXWOLPY-UHFFFAOYSA-N
Synonym 4-amino-n'-hydroxybenzimidamide, 4-amino-n'-hydroxybenzene-1-carboximidamide, z-4-amino-n'-hydroxybenzenecarboximidamide
PubChem CID 5478941
IUPAC Name 4-amino-N'-hydroxybenzenecarboximidamide
SMILES C1=CC(=CC=C1C(=NO)N)N

Specifications

Melting Point 168°C to 170°C
Quantity 1 g
Formula Weight 151.17
Percent Purity 97%
Chemical Name or Material 4-Aminobenzamidoxime
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only