missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzamidine Dihydrochloride 98.0+%, TCI America™

Catalog No. A21155G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21155G 5 g
A211525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21155G Supplier TCI America Supplier No. A21155G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2498-50-2
Molecular Formula C7H11Cl2N3
Molecular Weight (g/mol) 208.086
MDL Number MFCD00013001
InChI Key GHEHNICLPWTXJC-UHFFFAOYSA-N
Synonym 4-aminobenzamidine dihydrochloride, p-aminobenzamidine dihydrochloride, 4-aminobenzenecarboximidamide dihydrochloride, 4-aminobenzimidamide dihydrochloride, 4-aminobenzamidine 2hcl, 4-amidinoaniline dihydrochloride, benzenecarboximidamide, 4-amino-, dihydrochloride, unii-3c7vnu62ag, benzamidine, p-amino-, dihydrochloride, 3c7vnu62ag
PubChem CID 75626
ChEBI CHEBI:10618
IUPAC Name 4-aminobenzenecarboximidamide;dihydrochloride
SMILES C1=CC(=CC=C1C(=N)N)N.Cl.Cl

Specifications

Melting Point 291°C
Color White-Yellow
Quantity 5 g
Formula Weight 208.09
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminobenzamidine Dihydrochloride
RTECSNumber CV6126500
TSCA No
Recommended Storage Refrigerator