missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzamidine dihydrochloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 228050010

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC228050010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2498-50-2
208.09
GHEHNICLPWTXJC-UHFFFAOYSA-N
75626
4-aminobenzenecarboximidamide;dihydrochloride
C7H9N3·2HCl
MFCD00013001
4-aminobenzamidine dihydrochloride, p-aminobenzamidine dihydrochloride, 4-aminobenzenecarboximidamide dihydrochloride, 4-aminobenzimidamide dihydrochloride, 4-aminobenzamidine 2hcl, 4-amidinoaniline dihydrochloride, benzenecarboximidamide, 4-amino-, dihydrochloride, unii-3c7vnu62ag, benzamidine, p-amino-, dihydrochloride, 3c7vnu62ag
CHEBI:10618
C1=CC(=CC=C1C(=N)N)N.Cl.Cl

Specifications

4-Aminobenzamidine dihydrochloride
96.0
Authentic
Glass bottle
H2NC6H4C(=NH)NH2·2HCl
MFCD00013001
GHEHNICLPWTXJC-UHFFFAOYSA-N
4-aminobenzenecarboximidamide;dihydrochloride
75626
208.09
2498-50-2
100.0
96% min. (Argentometry)
C7H9N3·2HCl
1g
4-aminobenzamidine dihydrochloride, p-aminobenzamidine dihydrochloride, 4-aminobenzenecarboximidamide dihydrochloride, 4-aminobenzimidamide dihydrochloride, 4-aminobenzamidine 2hcl, 4-amidinoaniline dihydrochloride, benzenecarboximidamide, 4-amino-, dihydrochloride, unii-3c7vnu62ag, benzamidine, p-amino-, dihydrochloride, 3c7vnu62ag
C1=CC(=CC=C1C(=N)N)N.Cl.Cl
208.09
CHEBI:10618
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 219-692-2

SDS
Documents

Documents

Promotions

Promotions