missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoazobenzene 98.0+%, TCI America™

Catalog No. P014125G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P014125G 25 g
P0141100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P014125G Supplier TCI America Supplier No. P014125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60-09-3
Molecular Formula C12H11N3
Molecular Weight (g/mol) 197.241
MDL Number MFCD00007854
InChI Key QPQKUYVSJWQSDY-UHFFFAOYSA-N
Synonym 4-aminoazobenzene, p-aminoazobenzene, aniline yellow, 4-phenylazo aniline, cellitazol r, aminoazobenzene, 4-phenylazoaniline, benzenamine, 4-phenylazo, organol yellow, induline r
PubChem CID 6051
IUPAC Name 4-phenyldiazenylaniline
SMILES C1=CC=C(C=C1)N=NC2=CC=C(C=C2)N

Specifications

Melting Point 125°C
Quantity 25 g
UN Number 3077
Formula Weight 197.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminoazobenzene
EINECSNumber (3)-0363
RTECSNumber BY8225000
TSCA Yes