missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminoantipyrine, ≥98.0%, MilliporeSigma™ Supelco™
Specifications
Specifications
| Melting Point | 105°C to 110°C; 108°C to 110°C |
| CAS | 83-07-8 |
| Linear Formula | C11H13N3 O |
| Quantity | 25 g |
| Solubility Information | Water: 0.1 g/mL, clear |
| Formula Weight | 203.24 |
| Percent Purity | ≥98.0% (NT); ≥98.0% |
Safety and Handling
| RTECSNumber | CD2480000 |