missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoantipyrine, 97%

Catalog No. p-7021492
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1384622 100 g
AAA1384636 500 g
AAA138460E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1384622 Supplier Thermo Scientific Chemicals Supplier No. A1384622
Only null left

CAS: 83-07-8 | C11H13N3O | 203.245 g/mol

4-Aminoantipyrine shows both analgesic and anti-inflammatory properties. It is used as a reagent for glucose determination in the presence of peroxidase and phenol. It is used as a drug dipyrone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminoantipyrine shows both analgesic and anti-inflammatory properties. It is used as a reagent for glucose determination in the presence of peroxidase and phenol. It is used as a drug dipyrone.

Solubility
Soluble in water. 500 g/L (20°C), methanol. Partially soluble in diethyl ether.

Notes
Incompatible with Strong oxidizing agents, strong acids, acid chlorides, acid anhydrides.

Chemical Identifiers

CAS 83-07-8
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.245
MDL Number MFCD00003145
InChI Key RLFWWDJHLFCNIJ-UHFFFAOYSA-N
Synonym 4-aminoantipyrine, ampyrone, metapirazone, aminoantipyrine, solvapyrin-a, aminoazophene, 4-aminophenazone, 4-aminoantipyrene, aminoantipyrin, solnapyrin-a
PubChem CID 2151
ChEBI CHEBI:59026
IUPAC Name 4-amino-1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N

Specifications

Melting Point 106°C to 110°C
Assay Percent Range 97%
Quantity 100 g
Beilstein 181635
Merck Index 14,591
Solubility Information Soluble in water. 500g/L (20°C),methanol. Partially soluble in diethyl ether.
Formula Weight 203.25
Percent Purity 97%
Chemical Name or Material 4-Aminoantipyrine
Hazard Category H302-H315-H319-H335-H500
Hazard Statement GHS H Statement
H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 201-452-3
RTECSNumber CD2480000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only