missing translation for 'onlineSavingsMsg'
Learn More

4'-Aminoacetophenone 98.0+%, TCI America™

Catalog No. p-7116790
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A025125G 25 g
A0251100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A025125G Supplier TCI America Supplier No. A025125G
Only null left

Chemical Identifiers

CAS 99-92-3
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00007896
InChI Key GPRYKVSEZCQIHD-UHFFFAOYSA-N
Synonym 4'-aminoacetophenone, 4-aminoacetophenone, 1-4-aminophenyl ethanone, p-aminoacetophenone, 4-acetylaniline, p-acetylaniline, ethanone, 1-4-aminophenyl, p-aminoacetylbenzene, 1-4-aminophenyl ethan-1-one, acetophenone, p-amino
PubChem CID 7468
IUPAC Name 1-(4-aminophenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 107°C
Color Yellow
Boiling Point 293°C
Quantity 25 g
Formula Weight 135.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Aminoacetophenone
RTECSNumber AM5500000
TSCA Yes