missing translation for 'onlineSavingsMsg'
Learn More

4'-Aminoacetophenone, 99%

Catalog No. AC103090100
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103090100 10 g Glass bottle
AC103090250 25 g Glass bottle
AC103091000 100 g Glass bottle
AC103095000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC103090100 Supplier Thermo Scientific Chemicals Supplier No. 103090100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 99-92-3 | C8H9NO | 135.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-92-3
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00007896
InChI Key GPRYKVSEZCQIHD-UHFFFAOYSA-N
Synonym 4'-aminoacetophenone, 4-aminoacetophenone, 1-4-aminophenyl ethanone, p-aminoacetophenone, 4-acetylaniline, p-acetylaniline, ethanone, 1-4-aminophenyl, p-aminoacetylbenzene, 1-4-aminophenyl ethan-1-one, acetophenone, p-amino
PubChem CID 7468
IUPAC Name 1-(4-aminophenyl)ethanone
SMILES CC(=O)C1=CC=C(N)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 96.0°C to 102.0°C
Color Brown to Yellow
Boiling Point 293.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NC6H4COCH3
Quantity 10 g
Beilstein 14, 46
Fieser 04,18
Merck Index 14, 413
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in hcl,soluble in alcohol and ether,sparingly soluble in benzene
Formula Weight 135.17
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4'-Aminoacetophenone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-801-2

RUO – Research Use Only