missing translation for 'onlineSavingsMsg'
Learn More

4'-Aminoacetophenone, 99%

CAS: 99-92-3 | C8H9NO | 135.17 g/mol

Supplier:  Thermo Scientific Chemicals A1321318

 View more versions of this product

Catalog No. AAA1321318


Only null left
Explore available promotions

Description

Description

4'-Aminoacetophenone, is used in pharmaceuticals, medicines and other organic products. It is also used as a reagent for the photometric determination of Ce. As pharmaceutical intemediate or in the determination of palladium and vitaminB1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Aminoacetophenone, is used in pharmaceuticals, medicines and other organic products. It is also used as a reagent for the photometric determination of Ce. As pharmaceutical intemediate or in the determination of palladium and vitaminB1.

Solubility
It is soluble in hot water, ethanol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. acids, Incompatible to Acid chlorides, Acid anhydrides, Chloroformates, Strong oxidizing agents, Strong reducing agents. Stable under ordinary conditions.
Specifications

Specifications

4'-Aminoacetophenone
103°C to 108°C
C8H9NO
50 g
14,413
It is soluble in hot water,ethanol and ether.
CC(=O)C1=CC=C(N)C=C1
135.17
135.17
99-92-3
292°C to 294°C
MFCD00007896
471493
4'-aminoacetophenone, 4-aminoacetophenone, 1-4-aminophenyl ethanone, p-aminoacetophenone, 4-acetylaniline, p-acetylaniline, ethanone, 1-4-aminophenyl, p-aminoacetylbenzene, 1-4-aminophenyl ethan-1-one, acetophenone, p-amino
GPRYKVSEZCQIHD-UHFFFAOYSA-N
1-(4-aminophenyl)ethanone
7468
99%

Chemical Identifiers

99-92-3
135.17
GPRYKVSEZCQIHD-UHFFFAOYSA-N
7468
CC(=O)C1=CC=C(N)C=C1
C8H9NO
MFCD00007896
4'-aminoacetophenone, 4-aminoacetophenone, 1-4-aminophenyl ethanone, p-aminoacetophenone, 4-acetylaniline, p-acetylaniline, ethanone, 1-4-aminophenyl, p-aminoacetylbenzene, 1-4-aminophenyl ethan-1-one, acetophenone, p-amino
1-(4-aminophenyl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H312-H315-H319-H332-H335

EINECSNumber : 202-801-2

RTECSNumber : AM5500000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only