missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-N-Boc-L-phenylalanine, 95%

Catalog No. AAH5198003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5198003 1 g
AAH5198006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5198003 Supplier Thermo Scientific Chemicals Supplier No. H5198003
Only null left

CAS: 55533-24-9 | C14H20N2O4 | 280.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55533-24-9
Molecular Formula C14H20N2O4
Molecular Weight (g/mol) 280.32
MDL Number MFCD00038139
InChI Key NDMVQEZKACRLDP-NSHDSACASA-N
Synonym boc-phe 4-nh2-oh, boc-4-amino-l-phenylalanine, boc-l-4-aminophenylalanine, 4-amino-n-boc-l-phenylalanine, s-3-4-aminophenyl-2-tert-butoxycarbonyl amino propanoic acid, 4-amino-n-tert-butoxycarbonyl-l-phenylalanine, 2s-3-4-aminophenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-3-4-aminophenyl-2-tert-butoxycarbonylamino propanoic acid, 2s-3-4-aminophenyl-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid
PubChem CID 2761482
IUPAC Name (2S)-3-(4-aminophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(N)C=C1)C(O)=O

Specifications

Melting Point 124°C to 126°C
Quantity 1 g
Formula Weight 280.32
Percent Purity 95%
Chemical Name or Material 4-Amino-N-Boc-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only