missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-Amino-DL-phenylalanine

Catalog No. p-7058248
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6533103 1 g
AAJ6533106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6533103 Supplier Thermo Scientific Chemicals Supplier No. J6533103

CAS: 2922-41-0 | C9H12N2O2 | 180.207 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2922-41-0
Molecular Formula C9H12N2O2
Molecular Weight (g/mol) 180.207
MDL Number MFCD00007917
InChI Key CMUHFUGDYMFHEI-UHFFFAOYSA-N
Synonym 4-amino-dl-phenylalanine, 2-amino-3-4-aminophenyl propanoic acid, p-amino-dl-phenylalanine, p-aminophenylalanine, phenylalanine, 4-amino, 4-aminophenylalanine, h-dl-phe 4-nh2-oh, p-aminophenyl alanine, p-amino-dl-phenylalanine hydrate, l-2-amino-3-4-aminophenyl propanoic acid
PubChem CID 95174
IUPAC Name 2-amino-3-(4-aminophenyl)propanoic acid
SMILES C1=CC(=CC=C1CC(C(=O)O)N)N

Specifications

Melting Point 228°C to 230°C
Color White to Brown
Solubility Information Soluble in water
Quantity 1 g
Formula Weight 180.21
Physical Form Powder
Chemical Name or Material 4-Amino-DL-phenylalanine
EINECSNumber 220-869-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only