missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-methoxypyrimidine, 97%

CAS: 696-45-7 | C5H7N3O | 125.13 g/mol

$163.69 - $163.69

Chemical Identifiers

CAS 696-45-7
Molecular Formula C5H7N3O
Molecular Weight (g/mol) 125.13
MDL Number MFCD00129983
InChI Key VELRBZDRGTVGGT-UHFFFAOYSA-N
Synonym 4-amino-6-methoxypyrimidine, 6-methoxy-pyrimidin-4-ylamine, 6-methoxy-4-pyrimidinamine, 4-amino-6-methoxy pyrimidine, 4-pyrimidinamine, 6-methoxy, 6-methoxypyrimidine-4-ylamine, 6-methoxypyrimidin-4-amin, pubchem2605, acmc-209oa2, 6-amino-4-methoxypyrimidine
PubChem CID 735731
IUPAC Name 6-methoxypyrimidin-4-amine
SMILES COC1=NC=NC(=C1)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC337450050
SDS View Documents
Thermo Scientific Chemicals
337450050
5 g Glass Bottle
Each for $163.69
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

696-45-7
125.13
VELRBZDRGTVGGT-UHFFFAOYSA-N
735731
COC1=NC=NC(=C1)N
C5H7N3O
MFCD00129983
4-amino-6-methoxypyrimidine, 6-methoxy-pyrimidin-4-ylamine, 6-methoxy-4-pyrimidinamine, 4-amino-6-methoxy pyrimidine, 4-pyrimidinamine, 6-methoxy, 6-methoxypyrimidine-4-ylamine, 6-methoxypyrimidin-4-amin, pubchem2605, acmc-209oa2, 6-amino-4-methoxypyrimidine
6-methoxypyrimidin-4-amine

Specifications

696-45-7
100.0
White to Yellow
97%
C5H7N3O
5 g
VELRBZDRGTVGGT-UHFFFAOYSA-N
6-methoxypyrimidin-4-amine
735731
97%
4-Amino-6-methoxypyrimidine
96.0
154.0°C to 159.0°C
Authentic
Glass Bottle
MFCD00129983
4-amino-6-methoxypyrimidine, 6-methoxy-pyrimidin-4-ylamine, 6-methoxy-4-pyrimidinamine, 4-amino-6-methoxy pyrimidine, 4-pyrimidinamine, 6-methoxy, 6-methoxypyrimidine-4-ylamine, 6-methoxypyrimidin-4-amin, pubchem2605, acmc-209oa2, 6-amino-4-methoxypyrimidine
COC1=NC=NC(=C1)N
125.13
125.13
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only