missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-hydroxypyrazolo[3,4-d]pyrimidine 93.0+%, TCI America™

Catalog No. a2699100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2699100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2699100MG Supplier TCI America Supplier No. A2699100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5472-41-3
Molecular Formula C5H5N5O
Molecular Weight (g/mol) 151.129
MDL Number MFCD00005689
InChI Key KTQYLKORCCNJTQ-UHFFFAOYSA-N
Synonym 4-amino-6-hydroxypyrazolo 3,4-d pyrimidine, 4-amino-1h-pyrazolo 3,4-d pyrimidin-6-ol, 4-aminopyrazolo 3,4-d pyrimidin-6-ol, 4-amino-2h-pyrazolo 3,4-d pyrimidin-6-ol, 4-amino-1h-pyrazolo 3,4-d pyrimidin-6 7h-one, 4-aminopyrazolo 5,4-d pyrimidin-6-ol, acmc-1akdw, 4-amino-6-hydroxypyrazolo-3,4-d pyrimidine, 4-amino-1h-pyrazolo 4,3-e pyrimidin-6-ol, 4-amino-7-hydroxypyrazolo 3,4-d pyrimidine
PubChem CID 79623
IUPAC Name 4-amino-1,2-dihydropyrazolo[3,4-d]pyrimidin-6-one
SMILES C1=C2C(=NC(=O)N=C2NN1)N

Specifications

Quantity 100 mg
Formula Weight 151.13
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-6-hydroxypyrazolo[3,4-d]pyrimidine