missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-chloropyrimidine 98.0+%, TCI America™

Catalog No. A21365G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21365G 5 g
A21361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21365G Supplier TCI America Supplier No. A21365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5305-59-9
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.547
MDL Number MFCD00053576
InChI Key DUKKRSPKJMHASP-UHFFFAOYSA-N
Synonym 4-amino-6-chloropyrimidine, 6-chloropyrimidin-4-ylamine, 6-chloro-pyrimidin-4-ylamine, 4-pyrimidinamine, 6-chloro, 6-chloro-4-pyrimidinamine, 6-amino-4-chloropyrimidine, 6-chloropyrimidine-4-ylamine, 6-chloro-4-aminopyrimidine, 6-chlorpyrimidin-4-amin, zlchem 630
PubChem CID 238012
IUPAC Name 6-chloropyrimidin-4-amine
SMILES C1=C(N=CN=C1Cl)N

Specifications

Melting Point 220°C
Color White-Yellow
Quantity 5 g
Formula Weight 129.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-6-chloropyrimidine
EINECSNumber (9)-2241
TSCA No