missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-6-chloro-2-(methylthio)pyrimidine 98.0+%, TCI America™

Catalog No. A28485G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A28485G Supplier TCI America Supplier No. A28485G

Specifications

CAS 1005-38-5
Melting Point 133°C
Color White-Yellow
Molecular Formula C5H6ClN3S
Quantity 5 g
MDL Number MFCD00006088
Synonym 4-amino-6-chloro-2-methylthio pyrimidine, 4-amino-6-chloro-2-methylmercaptopyrimidine, 4-pyrimidinamine, 6-chloro-2-methylthio, 6-chloro-2-methylsulfanyl pyrimidin-4-amine, 6-chloro-2-methylthio pyrimidin-4-amine, pyrimidine, 4-amino-6-chloro-2-methylthio, 4-amino-6-chloro-2-methylthiopyrimidine, 6-chloro-2-methylthiopyrimidin-4-ylamine, 6-amino-4-chloro-2-methylmercaptopyrimidine, 6-chloro-2-methylsulfanyl-pyrimidin-4-amine
InChI Key ISUXMAHVLFRZQU-UHFFFAOYSA-N
SMILES CSC1=NC(=CC(=N1)Cl)N
IUPAC Name 6-chloro-2-methylsulfanylpyrimidin-4-amine
Molecular Weight (g/mol) 175.634
PubChem CID 70496
Formula Weight 175.63
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-6-chloro-2-(methylthio)pyrimidine
Show More Show Less