missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-bromopyrimidine, 98%

Catalog No. AAH6433503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6433503 1 g
AAH6433506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6433503 Supplier Thermo Scientific Chemicals Supplier No. H6433503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1439-10-7 | C4H4BrN3 | 174.001 g/mol

It is an inhibitor of thymidine phosphorylase, an enzyme known to be over-expressed in several tumor types and to play a role in metastasis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an inhibitor of thymidine phosphorylase, an enzyme known to be over-expressed in several tumor types and to play a role in metastasis.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 1439-10-7
Molecular Formula C4H4BrN3
Molecular Weight (g/mol) 174.001
MDL Number MFCD00233962
InChI Key IIFAONYUCDAVGA-UHFFFAOYSA-N
PubChem CID 10261576
IUPAC Name 5-bromopyrimidin-4-amine
SMILES C1=C(C(=NC=N1)N)Br

Specifications

Melting Point 207°C to 211°C
Color Brown
Quantity 1 g
Beilstein 112648
Synonym 4-amino-5-bromopyrimidine, 5-bromo-pyrimidin-4-ylamine, 4-pyrimidinamine, 5-bromo, 4-pyrimidinamine 5-bromo, 5-bromopyrimidine-4-ylamine, pubchem14978, acmc-209crn, 4-pyrimidinamine,5-bromo, 4-amino-5-bromo-pyrimidine, pyrimidine, 4-amino-5-bromo
Solubility Information Slightly soluble in water.
Formula Weight 174
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Amino-5-bromopyrimidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only