missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-bromo-2-chloropyrimidine, 95%

Catalog No. p-7053767
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6142906 5 g
AAH6142914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6142906 Supplier Thermo Scientific Chemicals Supplier No. H6142906

CAS: 205672-25-9 | C4H3BrClN3 | 208.44 g/mol

4-Amino-5-bromo-2-chloropyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-5-bromo-2-chloropyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Storage temperature: 2-4°C. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 205672-25-9
Molecular Formula C4H3BrClN3
Molecular Weight (g/mol) 208.44
MDL Number MFCD07782040
InChI Key QOWALNIZDHZTSM-UHFFFAOYSA-N
Synonym 4-amino-5-bromo-2-chloropyrimidine, 4-pyrimidinamine, 5-bromo-2-chloro, 5-bromo-2-chloro-4-pyrimidinamine, 5-bromo-2-chloro-pyrimidin-4-ylamine, 5-bromo-2-chloropyrimidine-4-ylamine, pubchem18570, acmc-209fbz, 5-bromo-2-chloro-pyrimidin-4-amine, 5-bromo-2-chloropyrimidin-4-ylamine
PubChem CID 2763293
IUPAC Name 5-bromo-2-chloropyrimidin-4-amine
SMILES NC1=NC(Cl)=NC=C1Br

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 208.44
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Amino-5-bromo-2-chloropyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only