missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-4H-1,2,4-triazole, 99%

Catalog No. AC104700250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104700250 25 g Glass bottle
AC104701000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104700250 Supplier Thermo Scientific Chemicals Supplier No. 104700250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 584-13-4 | C2H4N4 | 84.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 584-13-4
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.08
MDL Number MFCD00003099
InChI Key FMCUPJKTGNBGEC-UHFFFAOYSA-N
Synonym 4-amino-1,2,4-triazole, 4-amino-4h-1,2,4-triazole, 4h-1,2,4-triazol-4-amine, 1-amino-1,3,4-triazole, 4h-1,2,4-triazole, 4-amino, 1-amino-1h-1,3,4-triazole, 4-amino-1,2,4 4h-triazole, 4h-1,2,4-triazol-4-ylamine, 1-amino-1,4-triazole, 4-amino-1,4-triazole
PubChem CID 11432
IUPAC Name 1,2,4-triazol-4-amine
SMILES C1=NN=CN1N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 84.0°C
Color Beige or White
Flash Point >149°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 26, IV, 40
Solubility Information Solubility in water: 810g/L (20°C) in water. Other solubilities: soluble in ethanol and dimethyl sulfoxyde,very little soluble in vegetal oils
Formula Weight 84.08
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-4H-1, 2, 4-triazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Suspected of causing cancer.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-533-5

RUO – Research Use Only