missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-penten-2-one, 96%

Catalog No. p-7044819
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0837903 1 g
AAL0837906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0837903 Supplier Thermo Scientific Chemicals Supplier No. L0837903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1118-66-7 | C5H9NO | 99.133 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1118-66-7
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD00043715
InChI Key OSLAYKKXCYSJSF-ONEGZZNKSA-N
Synonym acetylacetonamine, fluoral-p, 4-amino-3-penten-2-one, e-4-aminopent-3-en-2-one, 3-penten-2-one, 4-amino, 3e-4-aminopent-3-en-2-one, 4-aminopent-3-en-2-one, 4-amino-pent-3-en-2-one, e-4-amino-3-pentene-2-one, 2-acetyl-1-amino-1-methylethylene
PubChem CID 5367854
ChEBI CHEBI:51695
IUPAC Name (E)-4-aminopent-3-en-2-one
SMILES CC(=CC(=O)C)N

Specifications

Melting Point 36°C to 41°C
Boiling Point 209°C to 213°C
Quantity 1 g
Beilstein 1209344
Formula Weight 99.13
Percent Purity 96%
Chemical Name or Material 4-Amino-3-penten-2-one
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only