missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-nitropyridine 98.0+%, TCI America™

Catalog No. p-7117809
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22605G 5 g
A226025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22605G Supplier TCI America Supplier No. A22605G
Only null left

Chemical Identifiers

CAS 1681-37-4
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.114
MDL Number MFCD00160304
InChI Key IUPPEELMBOPLDJ-UHFFFAOYSA-N
Synonym 4-amino-3-nitropyridine, 4-pyridinamine, 3-nitro, 3-nitro-4-pyridinamine, 3-nitro-4-aminopyridine, 3-nitro-pyridin-4-ylamine, 3-nitro-4-pyridylamine, pubchem1277, acmc-209dxo, 3-nitro-4-amino pyridine, 3-nitro-4-pyridinamine #
PubChem CID 548803
IUPAC Name 3-nitropyridin-4-amine
SMILES C1=CN=CC(=C1N)[N+](=O)[O-]

Specifications

Melting Point 205°C
Color Yellow
Quantity 5 g
Formula Weight 139.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-nitropyridine