missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-nitrophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog No. AC444570050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444570050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC444570050 Supplier Thermo Scientific Chemicals Supplier No. 444570050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 833486-94-5 | Molecular Formula: C12H17BN2O4 | Molecular Weight: 264.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 833486-94-5
Molecular Formula C12H17BN2O4
Molecular Weight (g/mol) 264.09
MDL Number MFCD06795680
InChI Key QOYJKGGBFKVKDP-UHFFFAOYSA-N
Synonym 4-amino-3-nitrophenylboronic acid pinacol ester, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, benzenamine, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-amino-3-nitrophenylboronic acid, pinacol ester, 2-4-amino-3-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-nitro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, pubchem22339, acmc-209ps0, ksc912g7t, qoyjkggbfkvkdp-uhfffaoysa-n
PubChem CID 16217914
IUPAC Name 2-nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(N)C(=C1)[N+]([O-])=O

Specifications

Melting Point 174°C to 177°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 264.09
Percent Purity 97%
Chemical Name or Material 4-Amino-3-nitrophenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only