missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-nitrobenzonitrile 98.0+%, TCI America™

Catalog No. p-7117913
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24475G 5 g
A244725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24475G Supplier TCI America Supplier No. A24475G
Only null left

Chemical Identifiers

CAS 6393-40-4
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD00013373
InChI Key JAHADAZIDZMHOP-UHFFFAOYSA-N
Synonym 3-nitro-4-aminobenzonitrile, 4-cyano-2-nitroaniline, benzonitrile, 4-amino-3-nitro, 4-amino-3-nitro-benzonitrile, 2-nitro-4-cyanoaniline, 4-amino-3-nitrobenzenecarbonitrile, zlchem 228, pubchem4669, 4-ammo-3-nitrobenzonitrile
PubChem CID 595901
IUPAC Name 4-amino-3-nitrobenzonitrile
SMILES NC1=CC=C(C=C1[N+]([O-])=O)C#N

Specifications

Melting Point 162°C
Quantity 5 g
UN Number 3439
Formula Weight 163.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-nitrobenzonitrile
EINECSNumber (3)-1791
TSCA No