missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-hydroxybutyric acid

Catalog No. p-7043029
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6514506 5 g
AAJ6514514 25 g
AAJ6514522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6514506 Supplier Thermo Scientific Chemicals Supplier No. J6514506
Only null left

CAS: 924-49-2 | C4H9NO3 | 119.12 g/mol

Reduces uptake of iodine by the thyroid glands in animals. Antispasmodic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Reduces uptake of iodine by the thyroid glands in animals
  • Antispasmodic

Chemical Identifiers

CAS 924-49-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00008141
InChI Key YQGDEPYYFWUPGO-UHFFFAOYSA-N
Synonym 4-amino-3-hydroxybutyric acid, gabob, 3-hydroxy-gaba, dl-4-amino-3-hydroxybutyric acid, gabomade, gamibetal, gaboril, buksamin, gabimex, gaminal
PubChem CID 2149
ChEBI CHEBI:16080
IUPAC Name 4-amino-3-hydroxybutanoic acid
SMILES C(C(CN)O)C(=O)O

Specifications

Melting Point 223°C (decomposition)
Color White to Yellow
Beilstein 1721708
Merck Index 14,444
Quantity 5 g
Formula Weight 119.12
Physical Form Powder
Chemical Name or Material 4-Amino-3-hydroxybutyric acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 213-106-9
RTECSNumber ES7015000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only