missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-3-hydroxybutyric Acid 98.0+%, TCI America™
$32.72 - $82.79
Chemical Identifiers
| CAS | 924-49-2 |
|---|---|
| Molecular Formula | C4H9NO3 |
| Molecular Weight (g/mol) | 119.12 |
| MDL Number | MFCD00008141 |
| InChI Key | YQGDEPYYFWUPGO-UHFFFAOYSA-N |
| Synonym | 4-amino-3-hydroxybutyric acid, gabob, 3-hydroxy-gaba, dl-4-amino-3-hydroxybutyric acid, gabomade, gamibetal, gaboril, buksamin, gabimex, gaminal |
| PubChem CID | 2149 |
| ChEBI | CHEBI:16080 |
| IUPAC Name | 4-amino-3-hydroxybutanoic acid |
| SMILES | C(C(CN)O)C(=O)O |
Chemical Identifiers
| 924-49-2 | |
| 119.12 | |
| YQGDEPYYFWUPGO-UHFFFAOYSA-N | |
| 2149 | |
| 4-amino-3-hydroxybutanoic acid |
| C4H9NO3 | |
| MFCD00008141 | |
| 4-amino-3-hydroxybutyric acid, gabob, 3-hydroxy-gaba, dl-4-amino-3-hydroxybutyric acid, gabomade, gamibetal, gaboril, buksamin, gabimex, gaminal | |
| CHEBI:16080 | |
| C(C(CN)O)C(=O)O |
Specifications
| 218°C | |
| White | |
| MFCD00008141 | |
| YQGDEPYYFWUPGO-UHFFFAOYSA-N | |
| 4-amino-3-hydroxybutanoic acid | |
| 119.12 | |
| CHEBI:16080 | |
| ≥98.0% (T) | |
| 4-Amino-3-hydroxybutyric Acid |
| 924-49-2 | |
| C4H9NO3 | |
| 4-amino-3-hydroxybutyric acid, gabob, 3-hydroxy-gaba, dl-4-amino-3-hydroxybutyric acid, gabomade, gamibetal, gaboril, buksamin, gabimex, gaminal | |
| C(C(CN)O)C(=O)O | |
| 5 g | |
| 2149 | |
| 119.12 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-0093
RTECSNumber : ES7015000
TSCA : Yes