missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-fluoropyridine 98.0+%, TCI America™

Catalog No. p-7117874
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23831G 1 g
A23835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23831G Supplier TCI America Supplier No. A23831G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2247-88-3
Molecular Formula C5H5FN2
Molecular Weight (g/mol) 112.107
MDL Number MFCD00234072
InChI Key UFIKBUVVVGSMGW-UHFFFAOYSA-N
Synonym 4-amino-3-fluoropyridine, 3-fluoro-4-aminopyridine, 3-fluoro-pyridin-4-ylamine, 3-fluoro-4-pyridylamine, 3-fluoro-4-pyridinamine, 4-pyridinamine, 3-fluoro, pubchem1261, 3-amine-5-fluoropyridine, 3-fluoropyridine-4-amine, acmc-1ck3i
PubChem CID 817122
IUPAC Name 3-fluoropyridin-4-amine
SMILES C1=CN=CC(=C1N)F

Specifications

Melting Point 79°C
Quantity 1 g
Formula Weight 112.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-fluoropyridine