missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-chloropyridine 97.0+%, TCI America™

Catalog No. A27881G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27881G 1 g
A27885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27881G Supplier TCI America Supplier No. A27881G
Only null left

Chemical Identifiers

CAS 19798-77-7
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD03984473
InChI Key KJBKPRMEMJKXDV-UHFFFAOYSA-N
Synonym 4-amino-3-chloropyridine, 3-chloro-pyridin-4-ylamine, 3-chloro-4-pyridinamine, 3-chloro-4-aminopyridine, 3-chloro-4-pyridinylamine, 4-pyridinamine, 3-chloro, 3-chloro-4-pyridinamin, 3-chloro-4-pyridylamine, 4-amino-3-chloro-pyridine, pubchem6653
PubChem CID 581853
IUPAC Name 3-chloropyridin-4-amine
SMILES C1=CN=CC(=C1N)Cl

Specifications

Melting Point 60°C
Quantity 1 g
Formula Weight 128.56
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-3-chloropyridine