missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-3-chlorobenzonitrile 98.0+%, TCI America™
$124.89 - $124.89
Chemical Identifiers
| CAS | 21803-75-8 |
|---|---|
| Molecular Formula | C7H5ClN2 |
| Molecular Weight (g/mol) | 152.58 |
| MDL Number | MFCD00052917 |
| InChI Key | OREVCMGFYSUYPX-UHFFFAOYSA-N |
| Synonym | 3-chloro-4-aminobenzonitrile, 2-chloro-4-cyanoaniline, 3-chloro-4-aminbenzonitrile, 4-amino-3-chloro-benzonitrile, benzonitrile, 4-amino-3-chloro, 4-amino-3-chlorobenzenecarbonitrile, pubchem4625, 2-chloro-4-cyanoanilin, 2-chloro-4-cyano-aniline, acmc-1cs04 |
| PubChem CID | 519896 |
| IUPAC Name | 4-amino-3-chlorobenzonitrile |
| SMILES | NC1=CC=C(C=C1Cl)C#N |
Chemical Identifiers
| 21803-75-8 | |
| 152.58 | |
| OREVCMGFYSUYPX-UHFFFAOYSA-N | |
| 519896 | |
| NC1=CC=C(C=C1Cl)C#N |
| C7H5ClN2 | |
| MFCD00052917 | |
| 3-chloro-4-aminobenzonitrile, 2-chloro-4-cyanoaniline, 3-chloro-4-aminbenzonitrile, 4-amino-3-chloro-benzonitrile, benzonitrile, 4-amino-3-chloro, 4-amino-3-chlorobenzenecarbonitrile, pubchem4625, 2-chloro-4-cyanoanilin, 2-chloro-4-cyano-aniline, acmc-1cs04 | |
| 4-amino-3-chlorobenzonitrile |
Specifications
| 104°C | |
| 5 g | |
| C7H5ClN2 | |
| 3439 | |
| OREVCMGFYSUYPX-UHFFFAOYSA-N | |
| 4-amino-3-chlorobenzonitrile | |
| 519896 | |
| ≥98.0% (GC) |
| 21803-75-8 | |
| Crystal-Powder at 20°C | |
| MFCD00052917 | |
| 3-chloro-4-aminobenzonitrile, 2-chloro-4-cyanoaniline, 3-chloro-4-aminbenzonitrile, 4-amino-3-chloro-benzonitrile, benzonitrile, 4-amino-3-chloro, 4-amino-3-chlorobenzenecarbonitrile, pubchem4625, 2-chloro-4-cyanoanilin, 2-chloro-4-cyano-aniline, acmc-1cs04 | |
| NC1=CC=C(C=C1Cl)C#N | |
| 152.58 | |
| 152.58 | |
| 4-Amino-3-chlorobenzonitrile |
Safety and Handling
TSCA : No